2-Amino-6-cyanpyrazin 97%

BD158289-250MG BD158289-1G BD158289-5G
BLDPBD158289-250MGEA 49 EUR
BLDPBD158289-250MG BLDPBD158289-1G BLDPBD158289-5G
2-Amino-6-cyanpyrazin 97%
2-Amino-6-cyanopyrazine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₅H₄N₄
Molecular Weight: 120,1100 g/mol
Lagertemperatur: Ambient
MDL: MFCD10697805
CAS-Nummer: 59489-39-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR