5-Iodopyrazin-2-amine 95%

Synonyme: 2-Amino-5-iodopyrazine

BD158286-100G BD158286-1G BD158286-10G BD158286-5G BD158286-25G
BLDPBD158286-100GEA 693 EUR
BLDPBD158286-100G BLDPBD158286-1G BLDPBD158286-10G BLDPBD158286-5G BLDPBD158286-25G
5-Iodopyrazin-2-amine 95%
5-Iodopyrazin-2-amine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C4H4IN3
Molecular Weight: 221,0000 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD09909691
CAS-Nummer: 886860-50-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR