2,6-Naphthyridin-1(2H)-one 97%

BLDPBD158284-1GEA 446 EUR
BLDPBD158284-1G BLDPBD158284-100MG BLDPBD158284-250MG
2,6-Naphthyridin-1(2H)-one 97%
2,6-Naphthyridin-1(2H)-one

Storage condition: Inert atmosphere, Room Temperature

Formel: C₈H₆N₂O
Molecular Weight: 146,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06762736
CAS-Nummer: 80935-77-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR