2,4-Dichloropyrido[3,4-d]pyrimidine 96%

BD158274-5G BD158274-1G BD158274-100MG BD158274-250MG
BLDPBD158274-5GEA 1637 EUR
BLDPBD158274-5G BLDPBD158274-1G BLDPBD158274-100MG BLDPBD158274-250MG
2,4-Dichloropyrido[3,4-d]pyrimidine 96%
2,4-Dichloropyrido[3,4-d]pyrimidine

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₃Cl₂N₃
Molecular Weight: 200,025 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD10699461
CAS-Nummer: 908240-50-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR