2,2,2-Trifluoro-1-(4-methoxyphenyl)ethanamine hydrochloride 96%

BLDPBD158259-1GEA 608 EUR
BLDPBD158259-1G BLDPBD158259-250MG BLDPBD158259-100MG
2,2,2-Trifluoro-1-(4-methoxyphenyl)ethanamine hydrochloride 96%
2,2,2-Trifluoro-1-(4-methoxyphenyl)ethanamine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formel: C₉H₁₁ClF₃NO
Molecular Weight: 241,63 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11506002
CAS-Nummer: 65686-77-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR