2'-Methoxy-5'-(trifluormethyl)acetophenon 98%

Synonyme: 1-(2-Methoxy-5-(trifluoromethyl)phenyl)ethanone,1-(2-Methoxy-5-trifluoromethylphenyl)ethanone

BD158221-1G BD158221-5G BD158221-100MG BD158221-250MG
BLDPBD158221-1GEA 181 EUR
BLDPBD158221-1G BLDPBD158221-5G BLDPBD158221-100MG BLDPBD158221-250MG
2'-Methoxy-5'-(trifluormethyl)acetophenon 98%
2'-Methoxy-5'-(trifluormethyl)acetophenon

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₉F₃O₂
Molecular Weight: 218,17 g/mol
Lagertemperatur: Ambient
MDL: MFCD09832300
CAS-Nummer: 503464-99-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR