(2R,4R)-1-(tert-Butoxycarbonyl)-4-phenylpyrrolidine-2-carboxylic acid 97%

BD158196-25MG BD158196-50MG BD158196-100MG
BLDPBD158196-25MGEA 221 EUR
BLDPBD158196-25MG BLDPBD158196-50MG BLDPBD158196-100MG
(2R,4R)-1-(tert-Butoxycarbonyl)-4-phenylpyrrolidine-2-carboxylic acid 97%
(2R,4R)-1-(tert-Butoxycarbonyl)-4-phenylpyrrolidine-2-carboxylic acid

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₆H₂₁NO₄
Molecular Weight: 291,34 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD09952342
CAS-Nummer: 158567-91-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR