4-Chloro-1H-pyrazol-3-amine 95+%

BD158151-100MG BD158151-5G BD158151-1G BD158151-250MG
BLDPBD158151-100MGEA 78 EUR
BLDPBD158151-100MG BLDPBD158151-5G BLDPBD158151-1G BLDPBD158151-250MG
4-Chloro-1H-pyrazol-3-amine 95+%
4-Chloro-1H-pyrazol-3-amine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₃H₄ClN₃
Molecular Weight: 117,537 g/mol
Lagertemperatur: Ambient
MDL: MFCD03419764
CAS-Nummer: 54301-34-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR