6-Chloro-N-methyl-2-pyridinamine 98%

Synonyme: 6-Chloro-N-methylpyridin-2-amine

BD158145-250MG BD158145-10G BD158145-100MG BD158145-5G BD158145-1G BD158145-25G
BLDPBD158145-250MGEA 32 EUR
BLDPBD158145-250MG BLDPBD158145-10G BLDPBD158145-100MG BLDPBD158145-5G BLDPBD158145-1G BLDPBD158145-25G
6-Chloro-N-methyl-2-pyridinamine 98%
6-Chloro-N-methyl-2-pyridinamine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₆H₇ClN₂
Molecular Weight: 142,59 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD00234196
CAS-Nummer: 89026-78-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR