1,5-Dimethyl-1H-pyrazol-4-amine 95%

BD158129-5G BD158129-1G BD158129-250MG
BLDPBD158129-5GEA 262 EUR
BLDPBD158129-5G BLDPBD158129-1G BLDPBD158129-250MG
1,5-Dimethyl-1H-pyrazol-4-amine 95%
1,5-Dimethyl-1H-pyrazol-4-amine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₅H₉N₃
Molecular Weight: 111,14 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD02253774
CAS-Nummer: 121983-36-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR