5-Methoxypyridine-2-carbonitrile 98%

Synonyme: 5-Methoxypicolinonitrile,2-Cyano-5-methoxypyridine

BD158061-100MG BD158061-250MG BD158061-1G BD158061-5G
BLDPBD158061-100MGEA 24 EUR
BLDPBD158061-100MG BLDPBD158061-250MG BLDPBD158061-1G BLDPBD158061-5G
5-Methoxypyridine-2-carbonitrile 98%
5-Methoxypyridine-2-carbonitrile

Storage condition: Inert atmosphere, Room Temperature

Formel: C₇H₆N₂O
Molecular Weight: 134,135 g/mol
Lagertemperatur: Ambient
MDL: MFCD08543467
CAS-Nummer: 89809-63-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR