Methyl-3-oxocyclopentanecarboxylate 98%

Synonyme: Methyl 3-oxocyclopentanecarboxylate,Methyl 3-oxocyclopentane-1-carboxylate

BD158053-5G BD158053-100MG BD158053-1G BD158053-10G BD158053-250MG BD158053-25G
BLDPBD158053-5GEA 69 EUR
BLDPBD158053-5G BLDPBD158053-100MG BLDPBD158053-1G BLDPBD158053-10G BLDPBD158053-250MG BLDPBD158053-25G
Methyl-3-oxocyclopentanecarboxylate 98%
Methyl-3-oxocyclopentanecarboxylate

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₇H₁₀O₃
Molecular Weight: 142,1525 g/mol
Lagertemperatur: Ambient
MDL: MFCD09832106
CAS-Nummer: 32811-75-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR