1-(1-Bromoethyl)-2-fluorobenzene 97%

BD158048-1G BD158048-5G BD158048-100MG BD158048-250MG
BLDPBD158048-1GEA 283 EUR
BLDPBD158048-1G BLDPBD158048-5G BLDPBD158048-100MG BLDPBD158048-250MG
1-(1-Bromoethyl)-2-fluorobenzene 97%
1-(1-Bromoethyl)-2-fluorobenzene

Storage condition: Inert atmosphere, 2-8°C

Formel: C₈H₈BrF
Molecular Weight: 203,05 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD11180278
CAS-Nummer: 405931-46-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR