2-Bromo-6-isopropylpyridine 97%

BD158039-250MG BD158039-10G BD158039-1G BD158039-5G
BLDPBD158039-250MGEA 38 EUR
BLDPBD158039-250MG BLDPBD158039-10G BLDPBD158039-1G BLDPBD158039-5G
2-Bromo-6-isopropylpyridine 97%
2-Bromo-6-isopropylpyridine

Storage condition: Inert atmosphere, Room Temperature

Formel: C₈H₁₀BrN
Molecular Weight: 200,08 g/mol
Lagertemperatur: Ambient
MDL: MFCD11223220
CAS-Nummer: 1037223-35-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR