6-Chlor-5-nitrochinolin 98%

BD158037-1G BD158037-250MG BD158037-10G BD158037-100MG BD158037-25G BD158037-5G
BLDPBD158037-1GEA 17 EUR
BLDPBD158037-1G BLDPBD158037-250MG BLDPBD158037-10G BLDPBD158037-100MG BLDPBD158037-25G BLDPBD158037-5G
6-Chlor-5-nitrochinolin 98%
6-Chloro-5-nitroquinoline

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₅ClN₂O₂
Molecular Weight: 208,6012 g/mol
Lagertemperatur: Ambient
MDL: MFCD00093616
CAS-Nummer: 86984-32-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR