3-Iod-6-methoxy-1H-indazol 97%

BLDPBD158003-1GEA 144 EUR
BLDPBD158003-1G BLDPBD158003-100MG BLDPBD158003-250MG
3-Iod-6-methoxy-1H-indazol 97%
3-Iodo-6-methoxy-1H-indazole
Formel: C₈H₇IN₂O
Molecular Weight: 274,0585 g/mol
MDL: MFCD09266218
CAS-Nummer: 936138-17-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR