4-(4,4-Dimethylpiperidin-1-yl)benzoic acid 95%

BD157994-1G BD157994-100MG BD157994-250MG
BLDPBD157994-1GEA 241 EUR
BLDPBD157994-1G BLDPBD157994-100MG BLDPBD157994-250MG
4-(4,4-Dimethylpiperidin-1-yl)benzoic acid 95%
4-(4,4-Dimethylpiperidin-1-yl)benzoic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₄H₁₉NO₂
Molecular Weight: 233,3100 g/mol
Lagertemperatur: Ambient
MDL: MFCD09031689
CAS-Nummer: 406233-26-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR