2-(5-Bromopyrimidin-2-yl)acetic acid 95%

BD157940-1G BD157940-250MG
BLDPBD157940-1GEA 84 EUR
BLDPBD157940-1G BLDPBD157940-250MG
2-(5-Bromopyrimidin-2-yl)acetic acid 95%
2-(5-Bromopyrimidin-2-yl)acetic acid

Storage condition: Sealed in dry, Store in freezer, under -20°C

Formel: C₆H₅BrN₂O₂
Molecular Weight: 217,0201 g/mol
Lagertemperatur: Freezer
MDL: MFCD09909705
CAS-Nummer: 1134327-93-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR