4-Fluor-3-nitroanisol 98%

BLDPBD157879-100GEA 420 EUR
BLDPBD157879-0.025 BLDPBD157879-100G BLDPBD157879-5G BLDPBD157879-1G BLDPBD157879-500G BLDPBD157879-25G BLDPBD157879-10G BLDPBD157879-250MG
4-Fluor-3-nitroanisol 98%
4-Fluor-3-nitroanisol

Storage condition: Sealed in dry, Room Temperature

Formel: C₇H₆FNO₃
Molecular Weight: 171,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11848442
CAS-Nummer: 61324-93-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR