5-Brom-4-methyl-1H-indazol 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD157870-10GEA 278 EUR
BLDPBD157870-10G BLDPBD157870-1G BLDPBD157870-5G BLDPBD157870-100MG BLDPBD157870-250MG
5-Brom-4-methyl-1H-indazol 98%
5-Bromo-4-methyl-1H-indazole

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₇BrN₂
Molecular Weight: 211,0600 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11007842
CAS-Nummer: 1082041-34-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR