5-Brom-4-chlor-1H-indazol 98%

BD157868-10G BD157868-25G BD157868-250MG BD157868-1G BD157868-5G
BLDPBD157868-10GEA 246 EUR
BLDPBD157868-10G BLDPBD157868-25G BLDPBD157868-250MG BLDPBD157868-1G BLDPBD157868-5G
5-Brom-4-chlor-1H-indazol 98%
5-Bromo-4-chloro-1H-indazole

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₇H₄BrClN₂
Molecular Weight: 231,4771 g/mol
Lagertemperatur: Ambient
MDL: MFCD11007933
CAS-Nummer: 1082041-90-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR