Ethyl-5-brom-2-thiophencarboxylat 98%

Synonyme: Ethyl 5-bromothiophene-2-carboxylate

BD157858-1G BD157858-10G BD157858-100G BD157858-25G BD157858-5G
BLDPBD157858-1GEA 7 EUR
BLDPBD157858-1G BLDPBD157858-10G BLDPBD157858-100G BLDPBD157858-25G BLDPBD157858-5G
Ethyl-5-brom-2-thiophencarboxylat 98%
Ethyl-5-brom-2-thiophencarboxylat

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₇H₇BrO₂S
Molecular Weight: 235,10 g/mol
Lagertemperatur: Ambient
MDL: MFCD02683089
CAS-Nummer: 5751-83-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR