Methyl-3-amino-6-brom-2-pyrazincarboxylat 97%

Synonyme: Methyl 3-amino-6-bromopyrazine-2-carboxylate

BD157815-10G BD157815-500G BD157815-100G BD157815-5G BD157815-25G BD157815-1G
BLDPBD157815-10GEA 46 EUR
BLDPBD157815-10G BLDPBD157815-500G BLDPBD157815-100G BLDPBD157815-5G BLDPBD157815-25G BLDPBD157815-1G
Methyl-3-amino-6-brom-2-pyrazincarboxylat 97%
Methyl-3-amino-6-bromopyrazine-2-carboxylate

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₆H₆BrN₃O₂
Molecular Weight: 232,03 g/mol
Lagertemperatur: Ambient
MDL: MFCD00834964
CAS-Nummer: 6966-01-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR