4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-trityl-1H-pyrazole 97%

BD157800-5G BD157800-250MG BD157800-25G BD157800-1G BD157800-10G
BLDPBD157800-5GEA 214 EUR
BLDPBD157800-5G BLDPBD157800-250MG BLDPBD157800-25G BLDPBD157800-1G BLDPBD157800-10G
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-trityl-1H-pyrazole 97%
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-trityl-1H-pyrazole

Storage condition: Inert atmosphere, 2-8°C

Formel: C₂₈H₂₉BN₂O₂
Molecular Weight: 436,3531 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD10698507
CAS-Nummer: 863238-73-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR