7-Bromobenzo[d]thiazole 98%

BD157792-1G BD157792-5G BD157792-100MG BD157792-250MG
BLDPBD157792-1GEA 133 EUR
BLDPBD157792-1G BLDPBD157792-5G BLDPBD157792-100MG BLDPBD157792-250MG
7-Bromobenzo[d]thiazole 98%
7-Bromobenzo[d]thiazole

Storage condition: Sealed in dry, Room Temperature

Formel: C₇H₄BrNS
Molecular Weight: 214,0824 g/mol
Lagertemperatur: Ambient
MDL: MFCD11858589
CAS-Nummer: 767-70-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR