(4-Cyanphenyl)essigsäureethylester 98%

Synonyme: Ethyl-(4-cyanphenyl)acetat

BD157753-1G BD157753-25G BD157753-5G BD157753-10G
BLDPBD157753-1GEA 17 EUR
BLDPBD157753-1G BLDPBD157753-25G BLDPBD157753-5G BLDPBD157753-10G
(4-Cyanphenyl)essigsäureethylester 98%
(4-Cyanphenyl)essigsäureethylester

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₁H₁₁NO₂
Molecular Weight: 189,21 g/mol
Lagertemperatur: Ambient
MDL: MFCD09953133
CAS-Nummer: 1528-41-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR