9-Phenanthrencarbaldehyd 97%

Synonyme: 9-Phenanthrenaldehyd

BLDPBD15773-25GEA 495 EUR
BLDPBD15773-25G BLDPBD15773-1G BLDPBD15773-250MG BLDPBD15773-5G
9-Phenanthrencarbaldehyd 97%
9-Phenanthrencarbaldehyd

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₅H₁₀O
Molecular Weight: 206,24 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00001175
CAS-Nummer: 4707-71-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR