4-Bromo-1H-benzoimidazole 98%

BD157700-250MG BD157700-5G BD157700-1G
BLDPBD157700-250MGEA 30 EUR
BLDPBD157700-250MG BLDPBD157700-5G BLDPBD157700-1G
4-Bromo-1H-benzoimidazole 98%
4-Bromo-1H-benzoimidazole

Storage condition: Sealed in dry, Room Temperature

Formel: C₇H₅BrN₂
Molecular Weight: 197,0320 g/mol
Lagertemperatur: Ambient
MDL: MFCD09754164
CAS-Nummer: 83741-35-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR