(4-Carboxybutyl)triphenylphosphoniumbromid 98%

BD1577-5G BD1577-10G BD1577-1000G BD1577-100G BD1577-25G BD1577-500G
BLDPBD1577-5GEA 7 EUR
BLDPBD1577-5G BLDPBD1577-10G BLDPBD1577-1000G BLDPBD1577-100G BLDPBD1577-25G BLDPBD1577-500G
(4-Carboxybutyl)triphenylphosphoniumbromid 98%
(4-Carboxybutyl)triphenylphosphonium bromide

Storage condition: Inert atmosphere, Room Temperature

Formel: C₂₃H₂₄BrO₂P
Molecular Weight: 443,32 g/mol
Lagertemperatur: Ambient
MDL: MFCD00011906
CAS-Nummer: 17814-85-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR