5-Isobutylimidazolidine-2,4-dione 95%

BD157671-250MG
BLDPBD157671-250MGEA 16 EUR
BLDPBD157671-250MG
5-Isobutylimidazolidine-2,4-dione 95%
5-Isobutylimidazolidine-2,4-dione

Storage condition: Inert atmosphere, Room Temperature

Formel: C₇H₁₂N₂O₂
Molecular Weight: 156,19 g/mol
Lagertemperatur: Ambient
MDL: MFCD00238853
CAS-Nummer: 67337-73-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR