3'-Fluor-2'-hydroxyacetophenon 97%
|
|
BLDPBD157670-100MGEA
23
EUR
BLDPBD157670-100MG
BLDPBD157670-10G
BLDPBD157670-250MG
BLDPBD157670-1G
BLDPBD157670-25G
BLDPBD157670-5G
3'-Fluor-2'-hydroxyacetophenon 97%
3'-Fluor-2'-hydroxyacetophenon
Storage condition: Inert atmosphere, Room Temperature
Formel:
C₈H₇FO₂ Molecular Weight: 154,14 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09055140 CAS-Nummer: 699-92-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...