4,6-Dichloro-2-(chloromethyl)pyrimidine 97%

BLDPBD157628-1GEA 277 EUR
BLDPBD157628-1G
4,6-Dichloro-2-(chloromethyl)pyrimidine 97%
4,6-Dichloro-2-(chloromethyl)pyrimidine

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₅H₃Cl₃N₂
Molecular Weight: 197,44 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD10697352
CAS-Nummer: 19875-05-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR