4-Thiophen-2-ylphenol 98%

BLDPBD157621-250MGEA 37 EUR
BLDPBD157621-250MG BLDPBD157621-100MG
4-Thiophen-2-ylphenol 98%
4-Thiophen-2-ylphenol
Formel: C₁₀H₈OS
Molecular Weight: 176,235 g/mol
MDL: MFCD06797407
CAS-Nummer: 29886-65-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR