1-(4-Fluorphenyl)-1,3-dihydro-2-benzofuran-5-carbonitril 97%

BLDPBD15762-250MGEA 18 EUR
BLDPBD15762-250MG
1-(4-Fluorphenyl)-1,3-dihydro-2-benzofuran-5-carbonitril 97%
1-(4-Fluorophenyl)-1,3-dihydroisobenzofuran-5-carbonitrile
Formel: C₁₅H₁₀FNO
Molecular Weight: 239,25 g/mol
MDL: MFCD06200812
CAS-Nummer: 64169-67-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR