6-Fluoroquinoxalin-2(1H)-one 95%

BD157619-250MG
BLDPBD157619-250MGEA 73 EUR
BLDPBD157619-250MG
6-Fluoroquinoxalin-2(1H)-one 95%
6-Fluoroquinoxalin-2(1H)-one
Formel: C₈H₅FN₂O
Molecular Weight: 164,1365 g/mol
MDL: MFCD09834133
CAS-Nummer: 55687-23-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR