7-Chinolinamin 97%

BLDPBD157604-10GEA 187 EUR
BLDPBD157604-10G BLDPBD157604-5G BLDPBD157604-1G BLDPBD157604-100G BLDPBD157604-25G BLDPBD157604-250MG
7-Chinolinamin 97%
7-Aminoquinoline

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₉H₈N₂
Molecular Weight: 144,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01646243
CAS-Nummer: 580-19-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR