Benzyl-N-[1-carbamoyl-2-(4-hydroxyphenyl)ethyl]carbamate 98%

Synonyme: Z-Tyr-NH₂,(S)-Benzyl (1-amino-3-(4-hydroxyphenyl)-1-oxopropan-2-yl)carbamate

BD157564-250MG BD157564-5G BD157564-1G
BLDPBD157564-250MGEA 29 EUR
BLDPBD157564-250MG BLDPBD157564-5G BLDPBD157564-1G
Benzyl-N-[1-carbamoyl-2-(4-hydroxyphenyl)ethyl]carbamate 98%
Benzyl-N-[1-carbamoyl-2-(4-hydroxyphenyl)ethyl]carbamate

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₇H₁₈N₂O₄
Molecular Weight: 314,341 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD00136595
CAS-Nummer: 19898-39-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR