Acridine-9-carboxylic acid 97%

Synonyme: 9-Carboxyacridine

BD15745-5G BD15745-10G BD15745-250MG BD15745-1G BD15745-100MG BD15745-25G
BLDPBD15745-5GEA 110 EUR
BLDPBD15745-5G BLDPBD15745-10G BLDPBD15745-250MG BLDPBD15745-1G BLDPBD15745-100MG BLDPBD15745-25G
Acridine-9-carboxylic acid 97%
Acridine-9-carboxylic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₄H₉NO₂
Molecular Weight: 223,227 g/mol
Lagertemperatur: Ambient
MDL: MFCD00009734
CAS-Nummer: 5336-90-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR