4-(2-((7-Amino-2-(furan-2-yl)-[1,2,4]triazolo[1,5-a][1,3,5]triazin-5-yl)amino)ethyl)phenol 99+%

BLDPBD157377-100MGEA 470 EUR
BLDPBD157377-100MG BLDPBD157377-250MG
4-(2-((7-Amino-2-(furan-2-yl)-[1,2,4]triazolo[1,5-a][1,3,5]triazin-5-yl)amino)ethyl)phenol 99+%
4-(2-((7-Amino-2-(furan-2-yl)-[1,2,4]triazolo[1,5-a][1,3,5]triazin-5-yl)amino)ethyl)phenol

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₆H₁₅N₇O₂
Molecular Weight: 337,33 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00908394
CAS-Nummer: 139180-30-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR