Z-L-Lys-SBzl hydrochloride 97%

BLDPBD157376-100MGEA 172 EUR
BLDPBD157376-100MG BLDPBD157376-50MG BLDPBD157376-250MG
Z-L-Lys-SBzl hydrochloride 97%
Z-L-Lys-SBzl hydrochloride

Storage condition: Inert atmosphere, 2-8°C

Formel: C₂₁H₂₇ClN₂O₃S
Molecular Weight: 422,968 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00058089
CAS-Nummer: 69861-89-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR