2'-(Trifluormethyl)biphenyl-3-carbonsäure 97%

Synonyme: 2'-(Trifluoromethyl)-[1,1'-biphenyl]-3-carboxylic acid

BD15720-1G BD15720-250MG BD15720-100MG BD15720-5G
BLDPBD15720-1GEA 120 EUR
BLDPBD15720-1G BLDPBD15720-250MG BLDPBD15720-100MG BLDPBD15720-5G
2'-(Trifluormethyl)biphenyl-3-carbonsäure 97%
2'-(Trifluormethyl)biphenyl-3-carbonsäure

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₄H₉F₃O₂
Molecular Weight: 266,22 g/mol
Lagertemperatur: Ambient
MDL: MFCD03424602
CAS-Nummer: 168618-48-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR