Bis(8-quinolinolato)zinc(II) hydrate 98+%

BLDPBD157153-5GEA 65 EUR
BLDPBD157153-5G BLDPBD157153-25G BLDPBD157153-1G
Bis(8-quinolinolato)zinc(II) hydrate 98+%
Bis(8-quinolinolato)zinc(II) hydrate

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₈H₁₂N₂O₂Zn
Molecular Weight: 353,69 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00065291
CAS-Nummer: 13978-85-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR