Z-D-MEAla-OH 97%

BLDPBD156959-10GEA 41 EUR
BLDPBD156959-10G BLDPBD156959-250MG BLDPBD156959-1G BLDPBD156959-100G BLDPBD156959-25G BLDPBD156959-5G BLDPBD156959-100MG
Z-D-MEAla-OH 97%
Z-D-MEAla-OH

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₂H₁₅NO₄
Molecular Weight: 237,255 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00153387
CAS-Nummer: 68223-03-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR