N-Cbz-D-glutamine 98%

BLDPBD156931-100MGEA 6 EUR
BLDPBD156931-100MG BLDPBD156931-1G BLDPBD156931-25G BLDPBD156931-5G BLDPBD156931-250MG
N-Cbz-D-glutamine 98%
N-Cbz-D-glutamine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₃H₁₆N₂O₅
Molecular Weight: 280,28 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 13139-52-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR