Z-D-Asp(OBzl)-OH 95%

Synonyme: N-Benzyloxycarbonyl-D-aspartic acid 4-benzyl ester,Z-D-Asp(OBzl)

BD156919-1G BD156919-10G BD156919-5G BD156919-250MG
BLDPBD156919-1GEA 32 EUR
BLDPBD156919-1G BLDPBD156919-10G BLDPBD156919-5G BLDPBD156919-250MG
Z-D-Asp(OBzl)-OH 95%
Z-D-Asp(OBzl)-OH

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₉H₁₉NO₆
Molecular Weight: 357,362 g/mol
Lagertemperatur: Ambient
MDL: MFCD00237336
CAS-Nummer: 5241-62-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR