Z-D-Arg-OH·HCl 97%

BLDPBD156916-5GEA 30 EUR
BLDPBD156916-5G BLDPBD156916-100G BLDPBD156916-1G BLDPBD156916-25G
Z-D-Arg-OH·HCl 97%
Z-D-Arg-OH·HCl

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₄H₂₁ClN₄O₄
Molecular Weight: 344,796 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00063010
CAS-Nummer: 113712-05-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR