2-Fluor-5-(trifluormethyl)benzophenon 97%

Synonyme: (2-Fluoro-5-(trifluoromethyl)phenyl)(phenyl)methanone

BD15681-5G BD15681-1G BD15681-25G
BLDPBD15681-5GEA 33 EUR
BLDPBD15681-5G BLDPBD15681-1G BLDPBD15681-25G
2-Fluor-5-(trifluormethyl)benzophenon 97%
2-Fluor-5-(trifluormethyl)benzophenon

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₄H₈F₄O
Molecular Weight: 268,21 g/mol
Lagertemperatur: Ambient
MDL: MFCD00061254
CAS-Nummer: 199292-40-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR