4,4'-Dibrombenzil 97%

BLDPBD15672-25GEA 136 EUR
BLDPBD15672-25G BLDPBD15672-100G BLDPBD15672-5G BLDPBD15672-10G BLDPBD15672-1G
4,4'-Dibrombenzil 97%
4,4'-Dibrombenzil

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₄H₈Br₂O₂
Molecular Weight: 368,02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00000104
CAS-Nummer: 35578-47-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR