(±)-δ-Tetradecalacton 98+%

Synonyme: (±)-5-Hydroxytetradecansäurelacton(±)-Tetradecan-5-olid(±)-Tetrahydro-6-nonyl-2H-pyran-2-on(±)-δ-Tetradekalacton

BLDPBD15653-25GEA 24 EUR
BLDPBD15653-25G BLDPBD15653-5G BLDPBD15653-500G BLDPBD15653-100G
(±)-δ-Tetradecalacton 98+%
(±)-δ-Tetradecalacton

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₄H₂₆O₂
Molecular Weight: 226,36 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00059809
CAS-Nummer: 2721-22-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR