(1R,5R)-4,6,6-Trimethylbicyclo[3.1.1]hept-3-en-2-one 97%

BD156404-100MG BD156404-1G BD156404-250MG
BLDPBD156404-100MGEA 80 EUR
BLDPBD156404-100MG BLDPBD156404-1G BLDPBD156404-250MG
(1R,5R)-4,6,6-Trimethylbicyclo[3.1.1]hept-3-en-2-one 97%
(1R,5R)-4,6,6-Trimethylbicyclo[3.1.1]hept-3-en-2-one

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₀H₁₄O
Molecular Weight: 150,21 g/mol
Lagertemperatur: Ambient
MDL: MFCD00064898
CAS-Nummer: 18309-32-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR